All Photos(1)

442537

Supelco

Decachlorobiphenyl

analytical standard

Empirical Formula (Hill Notation):
C12Cl10
CAS Number:
Molecular Weight:
498.66
Beilstein:
2065289
Ballschmiter Number:
209
EC Number:
MDL number:
PubChem Substance ID:

Quality Level

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

Clc1c(Cl)c(Cl)c(c(Cl)c1Cl)-c2c(Cl)c(Cl)c(Cl)c(Cl)c2Cl

InChI

1S/C12Cl10/c13-3-1(4(14)8(18)11(21)7(3)17)2-5(15)9(19)12(22)10(20)6(2)16

InChI key

ONXPZLFXDMAPRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product has been discontinued and product 31092-20mg may be an acceptable replacement.

Certificate of Analysis

Certificate of Origin

New strategy for comprehensive analysis of polybrominated diphenyl ethers, polychlorinated dibenzo-p-dioxins, polychlorinated dibenzofurans and polychlorinated biphenyls by gas chromatography coupled with mass spectrometry
Pirard C, et al.
Journal of Chromatography A, 998(1-2), 169-181 (2003)
Manual on Hydrocarbon Analysis null
A morphologic and physiologic bioassay for quantifying alterations in the lung following experimental chronic inhalation of tobacco smoke.
G L Huber et al.
Bulletin europeen de physiopathologie respiratoire, 17(2), 269-327 (1981-01-01)
Perchlorination reaction applied to the rapid determination of PCBs.
K Takamiya
Bulletin of environmental contamination and toxicology, 30(5), 600-605 (1983-05-01)
Xing Han et al.
Toxicology, 255(3), 177-186 (2008-11-22)
2,2',3,3',4,4',5,5',6,6'-Decachlorobiphenyl (PCB 209) is a fully chlorinated, non-coplanar biphenyl. To demonstrate that PCB 209 is not likely to exhibit human health hazards common to coplanar PCBs it was tested for cytochrome P450 (P450) enzyme induction potentials, genetic toxicity, and endocrine-modulating

Protocols

US EPA Method 8081: GC Analysis of Organochlorine Pesticides on Equity®-5

US EPA Method 8081: GC Analysis of Organochlorine Pesticides on Equity®-5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service