All Photos(1)

442873

Supelco

4-Nonylphenol

analytical standard

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
Beilstein:
2047450
EC Number:
MDL number:
PubChem Substance ID:

Quality Level

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

CCCCCCCCCc1ccc(O)cc1

InChI

1S/C15H24O/c1-2-3-4-5-6-7-8-9-14-10-12-15(16)13-11-14/h10-13,16H,2-9H2,1H3

InChI key

IGFHQQFPSIBGKE-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Product 442873 is being discontinued. Product 46405 is available and may be an acceptable replacement.

Certificate of Analysis

Certificate of Origin

Urinary concentrations of bisphenol A and 4-nonylphenol in a human reference population.
Calafat MA, et al.
Environmental Health Perspectives, 113(4), 391-395 (2004)
A H Shanthanagouda et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 157(2), 162-171 (2012-11-28)
This study investigated the influence of two endocrine disrupting chemicals (EDCs)-an exogenous oestrogen 17β-estradiol (E2) and the oestrogen mimic 4-n-nonylphenol (NP) on the expression of aromatase transcripts in both sexes of adult Murray river rainbowfish. Reproductively active mature fish were
Determination of the xenoestrogens 4-nonylphenol and bisphenol A by high-performance liquid chromatography and fluorescence detection after derivatisation with dansyl chloride.
Naassner, et al.
Journal of Chromatography A, 945(1-2), 133-138 (2002)
Caixiang Zhang et al.
Journal of chromatography. A, 1230, 110-116 (2012-02-22)
4-Nonylphenols (4-NPs) are known endocrine disruptors and by-products of the microbial degradation of nonylphenol polyethoxylate surfactants. One of the challenges to understanding the toxic effects of nonylphenols is the large number of isomers that may exist in environmental samples. In
Fatemeh Navid et al.
The Journal of investigative dermatology, 133(12), 2763-2770 (2013-05-09)
UVR suppresses the immune system through the induction of regulatory T cells (Tregs). UVR-induced DNA damage has been recognized as the major molecular trigger involved, as reduction of DNA damage by enhanced repair prevents the compromise to the immune system

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service