44926-U

Supelco

ISO 17943 57 Component VOC Mix

certified reference material, 200 μg/mL each component in methanol

NACRES:
NA.24

Quality Level

grade

certified reference material
TraceCERT®

Agency/Method

ISO 17943

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL each component in methanol

Featured Industry

Environmental

format

multi-component solution

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    tert-Amyl methyl ether
    Benzene
    Bromobenzene
    Bromochloromethane
    Bromodichloromethane
    Bromoform
    tert-Butylbenzene
    Butylbenzene
    sec-Butylbenzene
    tert-Butyl ethyl ether
    Carbon tetrachloride
    Chlorobenzene
    Chloroform
    2-Chlorotoluene
    4-Chlorotoluene
    Cumene
    1,2-Dibromo-3-chloropropane
    Dibromochloromethane
    1,2-Dibromoethane
    Dibromomethane
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    1,1-Dichloroethane
    1,2-Dichloroethane
    1,1-Dichloroethylene
    cis-1,2-Dichloroethylene
    trans-1,2-Dichloroethylene
    Dichloromethane
    1,2-Dichloropropane
    1,3-Dichloropropane
    2,2-Dichloropropane
    1,1-Dichloro-1-propene
    cis-1,3-Dichloropropene
    trans-1,3-Dichloropropene
    Ethylbenzene
    Hexachloro-1,3-butadiene
    p-Cymene
    Methyl tert-butyl ether
    Naphthalene
    Propylbenzene
    Styrene
    Toluene
    1,2,3-Trichlorobenzene
    1,2,4-Trichlorobenzene
    1,1,1,2-Tetrachloroethane
    1,1,2,2-Tetrachloroethane
    Tetrachloroethylene
    1,1,1-Trichloroethane
    1,1,2-Trichloroethane
    Trichloroethylene
    1,2,3-Trichloropropane
    1,2,4-Trimethylbenzene
    Mesitylene
    m-Xylene
    o-Xylene
    p-Xylene
    See All (57)

RIDADR

UN1230 - class 3 - PG 2 - Methanol, solution

Certificate of Analysis
Certificate of Origin
Protocols
High performance, reliability, and reproducibility of HS-SPME in combination with GC/MS for the determination of VOCs in water was proven in an interlaboratory trial. The new ISO 17943 using HS-SPME is an improvement on existing official methods for this determination in terms of sensitivity and selectivity.
Read More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.