47077
certified reference material
TraceCERT®
EPA 8240,8260
TraceCERT®
current certificate can be downloaded
ampule of 1 mL
25 μg/mL in methanol
HPLC: suitable
gas chromatography (GC): suitable
environmental
multi-component solution
2-8°C
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | 47414 | 47788 | Z569879 |
---|---|---|---|
grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® | grade analytical standard | grade - |
packaging ampule of 1 mL | packaging ampule of 1 mL, pkg of 4 ampules | packaging ampule of 1 mL | packaging - |
concentration 25 μg/mL in methanol | concentration 25 μg/mL in methanol | concentration in methyl tert-butyl ether (varied) | concentration - |
technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) - |
application(s) environmental | application(s) environmental | application(s) agriculture | application(s) - |
format multi-component solution | format multi-component solution | format multi-component solution | format - |
Danger
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
3 - Flammable liquids
WGK 2
49.5 °F - Solvent
9.7 °C - Solvent
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service