47207

Supelco

ASTM® D4815 Quantitative Calibration Mixture 3

with Internal Standard, (each component at the nominal wt% indicated), analytical standard

grade

analytical standard

Quality Level

Agency/Method

ASTM® D4815

packaging

ampule of 1 mL

concentration

(each component at the nominal wt% indicated)

application(s)

HPLC: suitable
gas chromatography (GC): suitable

Featured Industry

Petroleum

format

multi-component solution

storage temp.

room temp

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    tert-Butanol 5.7 % (w/w)
    tert-Butyl methyl ether 9.5 % (w/w)
    1,2-Dimethoxyethane 5 % (w/w)
    Ethanol 5.7 % (w/w)
    2-Methyl-2-butanol 2.38 % (w/w)
    Isooctane:xylene (65:35) 71.73 % (w/w)

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service