47758

Supelco

2-Bromo-1-chloropropane

analytical standard

Linear Formula:
CH3CH(Br)CH2Cl
CAS Number:
Molecular Weight:
157.44
EC Number:
MDL number:
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1000 mg

application(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.4783 (lit.)

bp

116-117 °C (lit.)

density

1.537 g/mL at 25 °C (lit.)

Featured Industry

Environmental

SMILES string

CC(Br)CCl

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Precautionary Statements

Target Organs

Respiratory system

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.