48152

Supelco

2,3,5,6-Tetrachlorophenol solution

certified reference material, 2000 μg/mL in methanol

CAS Number:
Beilstein/REAXYS Number:
2049586
MDL number:
PubChem Substance ID:

Quality Level

grade

certified reference material
TraceCERT®

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

application(s)

HPLC: suitable
gas chromatography (GC): suitable

Featured Industry

Environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Oc1c(Cl)c(Cl)cc(Cl)c1Cl

InChI

1S/C6H2Cl4O/c7-2-1-3(8)5(10)6(11)4(2)9/h1,11H

InChI key

KEWNKZNZRIAIAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

RIDADR

UN1230 - class 3 - PG 2 - Methanol

WGK Germany

2

Flash Point(F)

48.2 °F

Flash Point(C)

9 °C

Certificate of Analysis
Certificate of Origin
S Chauhan et al.
Applied microbiology and biotechnology, 52(2), 261-266 (1999-09-28)
Degradation of 2,4,5-trichlorophenol (2,4,5-TCP) and 2,3,5,6-tetrachlorophenol (TeCP) was studied using a two-stage approach that utilized efficient pulse electric discharge (PED) followed by biological degradation with a consortium from acclimated return activated sludge. The chlorinated phenols were treated in the PED...
Acute dermal toxicity of tetrachlorophenols in the rat.
S Y Shen et al.
Bulletin of environmental contamination and toxicology, 31(6), 680-685 (1983-12-01)
Articles
Separation of 2-Chlorophenol; 2,4-Dichlorophenol; 2,4,6-Tribromophenol; 2,4,6-Trichlorophenol; 2,4-Dinitrophenol; Pentafluorophenol; 2-Methylphenol, analytical standard; 2,3,4,6-Tetrachlorophenol; Pentachlorophenol; 4-Nitrophenol; 2-Bromophenol; 2,3,5,6-Tetrachlorophenol; 2,3,5-Trichlorophenol; 4-Chloro-3-methylphenol; 2,4,5-Trichlorophenol; 4-Methylphenol, analytical standard; 2,4-Dimethylphenol; 2-Nitrophenol; 3-Methylphenol, analytical standard; Phenol; 2-Methyl-4,6-dinitrophenol; 2,3,4-Trichlorophenol; 2,6-Dichlorophenol; 2,3,4,5-Tetrachlorophenol
Read More
Protocols
US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)
Read More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.