MilliporeSigma
All Photos(1)

48538

Supelco

Dichloromethane

analytical standard, stabilized

Synonym(s):
Methylene chloride
Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
EC Number:
MDL number:
PubChem Substance ID:

Quality Level

grade

analytical standard

Agency

OSHA 80

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

CofA

current certificate can be downloaded

autoignition temp.

1223 °F

quality

stabilized

expl. lim.

22 %

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−97 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

2-30°C

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

K E Mauck et al.
Integrative and comparative biology, 54(2), 193-209 (2014-04-22)
Recent research suggests that plant viruses, and other pathogens, frequently alter host-plant phenotypes in ways that facilitate transmission by arthropod vectors. However, many viruses infect multiple hosts, raising questions about whether these pathogens are capable of inducing transmission-facilitating phenotypes in
Angharad M Roberts et al.
Science translational medicine, 7(270), 270ra6-270ra6 (2015-01-16)
The recent discovery of heterozygous human mutations that truncate full-length titin (TTN, an abundant structural, sensory, and signaling filament in muscle) as a common cause of end-stage dilated cardiomyopathy (DCM) promises new prospects for improving heart failure management. However, realization
Silvia K Moreno-Forero et al.
The ISME journal, 9(1), 150-165 (2014-06-18)
The efficacy of inoculation of single pure bacterial cultures into complex microbiomes, for example, in order to achieve increased pollutant degradation rates in contaminated material (that is, bioaugmentation), has been frustrated by insufficient knowledge on the behaviour of the inoculated
Guilherme Asmar Alencar et al.
The Journal of clinical endocrinology and metabolism, 99(8), E1501-E1509 (2014-04-09)
Primary macronodular adrenal hyperplasia (PMAH) is a rare cause of Cushing's syndrome, usually characterized by functioning adrenal macronodules and increased cortisol production. Familial clustering of PMAH has been described, suggesting an inherited genetic cause for this condition. The aim of
Amélie Kochem et al.
Inorganic chemistry, 51(20), 10557-10571 (2012-09-28)
Square planar cobalt(II) complexes of salen ligands N,N'-bis(3-tert-butyl-5R-salicylidene)-1,2-cyclohexanediamine), where R = OMe (1) and tert-butyl (2), were prepared. 1 and 2 were electrochemically reversibly oxidized into cations [1-H(2)O](+) and [2-H(2)O](+) in CH(2)Cl(2). The chemically generated [1-H(2)O](SbF(6))·0.68 H(2)O·0.82CH(2)Cl(2) and [2-H(2)O](SbF(6))·0.3H(2)O·0.85CH(2)Cl(2) were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service