MilliporeSigma
All Photos(1)

48576

Supelco

Endosulfan I (alpha)

PESTANAL®, analytical standard

Synonym(s):
α-Endosulfan
Empirical Formula (Hill Notation):
C9H6Cl6O3S
CAS Number:
Molecular Weight:
406.93
Beilstein:
2950316
MDL number:
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

packaging

ampule of 25 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

room temp

SMILES string

ClC1=C(Cl)[C@@]2(Cl)[C@H]3COS(=O)OC[C@H]3[C@]1(Cl)C2(Cl)Cl

InChI

1S/C9H6Cl6O3S/c10-5-6(11)8(13)4-2-18-19(16)17-1-3(4)7(5,12)9(8,14)15/h3-4H,1-2H2/t3-,4+,7-,8+,19+

InChI key

RDYMFSUJUZBWLH-BEJSPLBDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service