4S6701-U

Supelco

EPA 8270/APP IX SemivolCalib Mix

certified reference material, 1000 μg/mL each component in methylene chloride

NACRES:
NA.24

Quality Level

grade

certified reference material
TraceCERT®

Agency/Method

EPA 8270

description

Separate Source

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 × 1 mL (4S6701-U)

concentration

1000 μg/mL each component in methylene chloride

application(s)

HPLC: suitable
gas chromatography (GC): suitable

Featured Industry

Environmental

format

multi-component solution

storage temp.

−20°C

General description

US EPA 8000 Series methods are analytical methods for identifying and quantifying halogenated and aromatic volatile organic compounds (VOCs) on the EPA′s Appendix VIII and Appendix IX lists in solid wastes and groundwater at active hazardous treatment, storage, and disposal sites. EPA method 8270 is typically used to determine semivolatile organic compounds.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    4-Aminobiphenyl
    Aniline
    Benzyl alcohol
    m-Cresol
    2,6-Dichlorophenol
    p-Dimethylaminoazobenzene
    7,12-Dimethylbenz[a]anthracene
    1,3-Dinitrobenzene
    Diphenylamine
    Ethyl methanesulfonate
    N-(2-Fluorenyl)acetamide
    Hexachloropropene
    Isosafrol
    Methapyrilene HCl
    3-Methylcholanthrene
    Methyl methanesulfonate
    2-Methyl-5-nitroaniline
    1-Naphthylamine
    2-Naphthylamine
    4-Nitroquinoline N-oxide
    N-Nitrosodibutylamine
    N-Nitrosodiethylamine
    N-Nitrosomethylethylamine
    Nitrosomorpholine
    1-Nitrosopiperidine
    1-Nitrosopyrrolidine
    Pentachlorobenzene
    Phenacetin
    Safrole
    1,2,4,5-Tetrachlorobenzene
    2,3,4,6-Tetrachlorophenol
    1,3,5-Trinitrobenzene
    See All (32)

Signal Word

Danger

Target Organs

Central nervous system

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 1593 6.1 / PGIII

WGK Germany

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.