50676-U

Hamilton PRP-C18 HPLC Column

5 μm particle size, L × I.D. 150 mm × 4.6 mm, stainless steel

NACRES:
SB.52

material

stainless steel

manufacturer/tradename

Hamilton

packaging

1 ea of

application(s)

HPLC: suitable

L × I.D.

150 mm × 4.6 mm

matrix

PS-DVB particle platform
polymeric

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

100 Å

operating pH

1-14

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

The Hamilton PRP-C18 HPLC column is designed to provide high-efficiency, reversed-phase separations over an extended column life in nearly any mobile phase or pH. The rigid stationary phase has excellent mechanical and thermal stability (> 100°C), does not shrink or swell, and is completely inert to most conditions commonly encountered in reversed-phase chromatography.

The PRP-C18 is a durable and effective column for general purpose HPLC and especially well-suited for specialized applications in the clinical, pharmaceutical, environmental, food, forensics, and life sciences industries. Benefits of the PRP-C18 HPLC Column:

  • Excellent mechanical and therma stability (> 100º C)
  • Can be regenerated in high acidic or basic concentrations (1 M)
  • High-efficiency reversed-phase separations in nearly any mobile phase or pH
  • Performs extreme applications not possible with traditional ODS C18

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service