507970

Supelco

JHAP-9 Mix

100 ppb in nitrogen, pkg of 110 L (pi-marked cylinder), analytical standard

EC Number:

grade

analytical standard

Quality Level

Agency/Method

EPA TO-14
JMHLW (Japan Ministry of Health and Welfare)

packaging

pkg of 110 L (pi-marked cylinder)

manufacturer/tradename

Scott Gas T9A10012-110

concentration

100 ppb in nitrogen

application(s)

HPLC: suitable
gas chromatography (GC): suitable

Featured Industry

Environmental

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Acrylonitrile
    Benzene
    1,3-Butadiene
    Chloroform
    1,2-Dichloroethane
    Dichloromethane
    Tetrachloroethylene
    Trichloroethylene
    Vinyl chloride
    See All (9)

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

RIDADR

UN1066 - class 2 - Nitrogen, compressed

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.