MilliporeSigma
All Photos(1)

51231-U

Upchurch Ultra-Low Volume Precolumn Filter

pore size 2 μm, pkg of 1 ea

NACRES:
SB.52

description

Solvent filter assembly with frit (1.4μL swept volume)

packaging

pkg of 1 ea

manufacturer/tradename

A-315 (OEM)

pore size

2 μm

Looking for similar products? Visit Product Comparison Guide

General description

These precolumn filters have a .020" diameter through hole, a stainless steel body, and are pressure rated to 9000 psi (620 bar). They will accept 1/16" tubing and standard 10-32 threaded high pressure fittings.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service