Documents

51233-U

Upchurch Ultra-Low Volume Precolumn Filter

pore size 0.5 μm, pkg of 1 ea

NACRES:
SB.52

description

solvent filter assembly (0.84 μL swept volume)

packaging

pkg of 1 ea

manufacturer/tradename

A-318 (OEM)

pore size

0.5 μm

General description

With a .010" diameter thru-hole, this filter has one of the lowest swept volumes of any HPLC precolumn filter available, ensuring maximum protection with very little band broadening. It is pressure rated to 9,000 psi (620 bar).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.