52220-U
Upchurch VHP (Very High Pressure) Fittings
VHP-920, for use with 1/32 inch (0.8mm) tubing, thread (M4 x 0.7), stainless steel
NACRES:
SB.52
material
stainless steel
description
Integrated ferrule
manufacturer/tradename
VHP-920
packaging
1 ea of
fitting
thread (M4 x 0.7)
compatibility
for use with 1/32 inch (0.8mm) tubing
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.