MilliporeSigma
All Photos(1)

55218-U

Supelco

SPE Frit Insertion Tool

for use with 3 mL SPE tubes

Synonym(s):
pk1 SPE Frit Insertion Tool, 3mL Tube
NACRES:
NB.21

material

polypropylene

Quality Level

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

compatibility

for use with 3 mL SPE tubes

Looking for similar products? Visit Product Comparison Guide

General description

Used to tighten or insert frits into SPE tubes.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service