Documents

55224-U

Supelco

SPE Frit Insertion Tool

for use with 20 and 60 mL SPE tubes

Synonym(s):
pk1 SPE Frit Insertion Tool, 20 & 60mL Tubes
NACRES:
NB.21
Pricing and availability is not currently available.

material

polypropylene material

Quality Level

Featured Industry

Food and Beverages

compatibility

for use with 20 and 60 mL SPE tubes

Looking for similar products? Visit Product Comparison Guide

General description

Used to tighten or insert frits into SPE tubes.
Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.