Documents

55710-U

Supelco

Discovery® C8 Validation Pack

5 μm particle size, L × I.D. 5 cm × 4.6 mm

Pricing and availability is not currently available.

Agency/Method

suitable for USP L7

packaging

pkg of 3 ea

application(s)

HPLC: suitable

L × I.D.

5 cm × 4.6 mm

matrix active group

C8 (octyl) phase

particle size

5 μm

pore size

180 Å

Featured Industry

Food and Beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Footnote

Pack includes 3 columns, each from a different lot of bonded phase.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Sigma-Aldrich Co. LLC
Certificate of Analysis
Certificate of Origin
Hugo M Botelho et al.
Scientific reports, 5, 9038-9038 (2015-03-13)
Plasma membrane proteins are essential molecules in the cell which mediate interactions with the exterior milieu, thus representing key drug targets for present pharma. Not surprisingly, protein traffic disorders include a large range of diseases sharing the common mechanism of...
Simeon Grazio et al.
Clinical and experimental rheumatology, 31(5), 665-671 (2013-06-07)
Using proteomic approach in this study, we sought to identify proteins with heparin affinity associated with rheumatoid arthritis (RA), psoriatic arthritis (PsA) and non-inflammatory arthritis (NIA). Plasma samples from adult RA, PsA and NIA patients, 20 of each, were collected....
G L Bundy et al.
The Journal of biological chemistry, 261(2), 747-751 (1986-01-15)
The gorgonian coral Pseudoplexaura porosa contains a lipoxygenase capable of converting exogenous arachidonic acid into (8R)-8-hydroperoxy-5,9,11,14-eicosatetraenoic acid. The (8R)- (or 8-L-) configuration in this product, opposite to that observed in previously reported 8-lipoxygenase products, was determined unambiguously by comparison of...
M Q Zhang et al.
Die Pharmazie, 46(10), 687-700 (1991-10-01)
The rapid growth in the quinolone research changed the whole face of the previous SAR concepts. So far structural modifications at all positions of the quinolone nucleus except the 4-oxo group have successfully lead to the discovery of potent antimicrobial...
Scott C Bell et al.
Pharmacology & therapeutics, 145, 19-34 (2014-06-17)
With the discovery of the CFTR gene in 1989, the search for therapies to improve the basic defects of cystic fibrosis (CF) commenced. Pharmacological manipulation provides the opportunity to enhance CF transmembrane conductance regulator (CFTR) protein synthesis and/or function. CFTR...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.