567222-U

Supelco

Discovery® BIO Wide Pore C8 HPLC Column

10 μm particle size, L × I.D. 25 cm × 10 mm

eCl@ss:
32110501
NACRES:
SB.52

Quality Level

Agency/Method

suitable for USP L7

feature

endcapped

manufacturer/tradename

Discovery

extent of labeling

5% Carbon loading

parameter

0-70 °C temperature
400 bar pressure (5801 psi)

application(s)

HPLC: suitable

L × I.D.

25 cm × 10 mm

surface coverage

4 μmol/m2

matrix

silica particle platform

matrix active group

C8 (octyl) phase

particle size

10 μm

pore size

300 Å

surface area

100 m2/g

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Discovery BIO Wide Pore C8 exhibits hydrophobicity intermediate between the Discovery BIO Wide Pore C5 and the Discovery BIO Wide Pore C18. The difference in hydrophobicity gives it unique selectivity relative to these other phases. It is ideal for peptide mapping because it provides complementary information compared to a C18 separation. Because of its intermediate hydrophobicity, we also recommend it for method development or scouting work. As with all Discovery BIO Wide Pore phases, the C8 phase gives efficient, symmetrical peaks, exceptional stability, long column lifetime, and LC-MS compatibility.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.