MilliporeSigma
  • 57043
All Photos(1)

57043

Supelco

Visiprep Manifold Plate (included with standard and DL models unless otherwise noted)

for use with 12-port model, 20 mL scintillation vials, not included with standard and DL models

Synonym(s):
20 ML SCINTILLATION VIAL PLATE

technique(s)

solid phase extraction (SPE): suitable

Quality Level

application(s)

food and beverages

compatibility

for use with 12-port model, 20 mL scintillation vials, not included with standard and DL models

Looking for similar products? Visit Product Comparison Guide

Packaging

1 ea

Legal Information

Visiprep is a trademark of Sigma-Aldrich Co. LLC

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service