Documents

575618-U

Supelco

Discovery® DSC-SAX SPE 96-well Plate

bed wt. 100 mg/well, pk of 1

Pricing and availability is not currently available.

composition

bed wt., 100 mg/well

packaging

pk of 1

application(s)

solid phase extraction (SPE): suitable

matrix active group

ammonium ion, quaternary bonding

Featured Industry

Food and Beverages

separation technique

ion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Retention Mechanism: Anion-exchange
Sample Matrix Compatibility: Organic or aqueous solutions

  • A polymerically bonded quarternary amine that remains positively charged at all pH levels
  • Counter ion is Cl-
  • Ion exchange capacity is ∼ 0.14 meq/g
  • Commonly used when extracting weaker anions (e.g., carboxylic acids) that may not bind strongly enough to weaker anion-exchangers
  • Selectivity can be modified by changing the counter ion with the appropriate buffer during conditioning

Legal Information

Discovery is a registered trademark of Sigma-Aldrich Co. LLC
Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.