577124-U

Supelco

Titan C18 UHPLC Column, 1.9 micron

1.9 μm particle size, L × I.D. 10 cm × 2.1 mm

NACRES:
SB.52

Quality Level

Agency/Method

suitable for USP L1

feature

endcapped

manufacturer/tradename

Titan

extent of labeling

13% carbon loading

parameter

18000 psi pressure (1241 bar)
60 °C max. temp.

application(s)

HPLC: suitable

L × I.D.

10 cm × 2.1 mm

matrix

monodisperse silica particle platform

matrix active group

C18 (octadecyl) phase

particle size

1.9 μm

pore size

0.76 cm3/g pore volume
80 Å pore size

surface area

410 m2/g

pH range

2 - 8

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Related Categories

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Titan is a trademark of Sigma-Aldrich Co. LLC

WGK Germany

WGK 1

  1. Which document(s) contains shelf-life or expiration date information for a given product?

    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

  2. How do I get lot-specific information or a Certificate of Analysis?

    The lot specific COA document can be found by entering the lot number above under the "Documents" section.

  3. How do I find price and availability?

    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

  4. What is the Department of Transportation shipping information for this product?

    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

  5. When are additional phases expected for Titan 1.9μm columns?

    Additional Titan 1.9μm phases are being developed and will encompass all aspects of UHPLC requirements.

  6. Are guard columns available for  Titan 1.9 μm C18 columns with 2.1 mm ID?

    Guard columns for Titan 1.9 μm C18 UHPLC columns with 2.1 mm ID are product 577127-U.  This product is a pack of three guard cartridges with dimensions 2.1 X 5 mm.   These cartridges require the use of the guard column holder, product 577133-U.

  7. Can I use Titan 1.9 μm columns on HPLC systems?

    Titan 1.9 μm columns are designed to be used only on UPLC or UHPLC systems.

  8. Which UHPLC Instruments can be used with Titan?

    Titan 1.9μm columns can be used on any UPLC or UHPLC System.

  9. What is the best practice for long-term storage of Titan C18 1.9 μm columns?

    Long-term storage of silica-based, reversed-phase columns is best in 100% acetonitrile. Columns may be safely stored for short periods (up to 3 or 4 days) in most common mobile phases. However, when using buffers, it is best to protect both the column and the HPLC equipment and remove any salts by flushing the column with the same mobile phase without the buffer (e.g., when using 60:40 ACN:buffer, flush the column with 60:40 ACN:H2O) to eliminate any danger of corrosion from the salts while providing rapid reequilibration of the column with the original mobile phase. Before storing the column, the end-fittings should be tightly sealed with the end-plugs that are provided with the column to prevent the packing from drying.

  10. What solvents are compatible with Titan C18 1.9 μm columns?

    Water and all common organic solvents are compatible with Titan C18 1.9 μm columns.

Fabrice Gritti et al.
Journal of chromatography. A, 1392, 10-19 (2015-03-24)
The kinetic performances of 3.0 × 100 mm columns packed with 1.9 μm Titan-C18 particles with average mesopore sizes of 80 Å and 120 Å were investigated quantitatively for the analysis of biomolecules. Large mesopores are expected to speed up...
Fabrice Gritti et al.
Journal of chromatography. A, 1355, 164-178 (2014-06-28)
In a previous report, it was reported that columns packed with fully porous 1.9μm Titan-C18 particles provided a minimum reduced plate height as small as 1.7 for the most retained compound (n-octanophenone) under RPLC conditions. These particles are characterized by...
Fabrice Gritti et al.
Journal of chromatography. A, 1355, 179-192 (2014-07-09)
The mass transfer mechanism in four prototype columns (2.1 and 3.0×50mm, 2.1 and 3.0×100mm) packed with 1.9μm fully porous Titan-C18 particles was investigated by using two previously reported home-made protocols. The first one was used to measure the eddy dispersion...
Fabrice Gritti et al.
Journal of chromatography. A, 1384, 76-87 (2015-02-11)
Previous data have shown that could deliver a minimum reduced plate height as small as 1.7. Additionally, the reduction of the mesopore size after C18 derivatization and the subsequent restriction for sample diffusivity across the Titan-C18 particles were found responsible...
Articles
Synthetic cannabinoids (e.g. "Spice") are a type of designer drug that provides a cannabis-type high.
Read More
Protocols
A comprehensive platform for lipidomics studies by LC/MS, applied without any modification in conditions and suitable to both ESI and APCI interfaces.
Read More
An MS-compatible alternative to the USP and EP methods, with improved specificity and reduced analysis time.
Read More
Immunoassay-based methods for digitoxin or digoxin exist, but are time consuming, labor intensive, and not selective due to similarities in their chemical structures.
Read More
SupelMIP SPE-Bisphenol A gave high recovery for baseline resolution of five related compounds with LC-Fluorescence detection.
Read More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.