Documents

58297C30

Supelco

SUPELCOSIL LC-8 HPLC Column

5 μm particle size, L × I.D. 25 cm × 3 mm

eCl@ss:
32110501
NACRES:
SB.52
Pricing and availability is not currently available.

Quality Level

Agency/Method

suitable for USP L7

feature

endcapped

mfr. no.

SUPELCOSIL

extent of labeling

6.0% carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

application(s)

HPLC: suitable

L × I.D.

25 cm × 3 mm

surface coverage

surface coverage 3.2 μmol/m2

matrix

silica gel, spherical particle platform

matrix active group

C8 (octyl) phase

particle size

5 μm

pore size

120 Å

surface area

170 m2/g

pH range

2 - 7.5

Featured Industry

Food and Beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

A phase less hydrophobic than C18. Provides less retention of both polar and non-polar compounds than C18. Use a mobile phase containing 5% less organic modifier for the C8 column than C18. Polar compounds are, relatively, more strongly retained on C8 than C18 columns.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC
Certificate of Analysis
Certificate of Origin
F Belal et al.
Journal of pharmaceutical and biomedical analysis, 24(3), 335-342 (2001-02-24)
A simple, rapid and sensitive HPLC method has been developed for the simultaneous determination of ramipril and hydrochlorothiazide in their dosage forms. Acetonitrile: sodium perchlorate solution (0.1 M) adjusted to pH 2.5+/-0.2 with phosphoric acid (46:54 v/v), was used as...
M Molíková et al.
Journal of chromatography. A, 1217(8), 1305-1312 (2010-01-12)
The availability of ionic liquids (ILs) in wide areas of application often results in the requirement on their determination. The attention is also often focused on the knowledge of hydrophobicity as it plays a key role in the biological effects...
J H Nichols et al.
Clinical chemistry, 40(7 Pt 1), 1312-1316 (1994-07-01)
This automated assay determines the concentration of the antidepressant fluoxetine (Prozac) and its active metabolite norfluoxetine in serum by reversed-phase HPLC with spectrophotometric detection. Extraction, injection, and quantification are performed by the Gilson Aspec automated sample handler. The patient's specimen...
Vesna Cerkvenik-Flajs et al.
Analytica chimica acta, 663(2), 165-171 (2010-03-09)
An analytical method has been developed for the simultaneous determination of the following endectocide drugs in milk: ivermectin, abamectin, doramectin, moxidectin, eprinomectin, emamectin and nemadectin. Samples were extracted with acetonitrile, purified with solid-phase extraction on a reversed phase C(8), derivatised...
K Ary et al.
Journal of pharmaceutical and biomedical analysis, 26(2), 179-187 (2001-07-27)
A reversed-phase high-performance liquid chromatographic method with coulometric and UV detection has been developed for the simultaneous determination of morphine, morphine-3-glucuronide and morphine-6-glucuronide. The separation was carried out by using a Supelcosil LC-8 DB reversed-phase column and 0.1 M potassium...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.