Documents

58625-U

Supelco

Cross

for connecting 1/16 in., bore size 0.25 mm

Synonym(s):
Valco Unions
NACRES:
SB.52
Pricing and availability is not currently available.

packaging

pkg of 1 ea

mfr. no.

VICI Valco®, ZX1C

bore size

0.25 mm

compatibility

for connecting 1/16 in.

Looking for similar products? Visit Product Comparison Guide

General description

Stainless steel, unless noted otherwise. Nuts and ferrules included.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Valco is a registered trademark of Valco Instruments Co. Inc.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.