Documents

59138C30

Supelco

SUPELCOSIL SAX1 HPLC Column

5 μm particle size, L × I.D. 25 cm × 3 mm

eCl@ss:
32110501
NACRES:
SB.52

Quality Level

Agency/Method

suitable for USP L14

manufacturer/tradename

SUPELCOSIL

extent of labeling

12% carbon loading

parameter

0-70 °C temperature
400 bar pressure (5801 psi)

application(s)

HPLC: suitable

L × I.D.

25 cm × 3 mm

matrix

silica gel, spherical particle platform

matrix active group

propyltrimethylammonium phase

particle size

5 μm

pore size

120 Å

surface area

170 m2/g

Featured Industry

Food and Beverages

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The SUPELCOSIL SAX1 column has a strongly basic quaternary aminopropyl phase and is used for separating anions.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.