MilliporeSigma
All Photos(1)

59485-U

Opti-Max® outlet check valve

for use with M45, M45G, M501, 510, 515, 590, 600, 600E, 610, 6K, 6KA

Synonym(s):
Optimize Technologies HPLC Pump Part Waters
NACRES:
SB.51

packaging

pkg of 1 ea

manufacturer/tradename

Waters 25216

compatibility

for use with M45, M45G, M501, 510, 515, 590, 600, 600E, 610, 6K, 6KA

Looking for similar products? Visit Product Comparison Guide

Legal Information

Opti-Max is a registered trademark of Optimize Technologies, Inc.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service