59515

Supelco

SUPELCOSIL LC-NH2-NP Supelguard Cartridge

5 μm particle size, L × I.D. 2 cm × 4 mm

EC Number:
eCl@ss:
32110501

Agency/Method

suitable for USP L8

packaging

pkg of 1 ea

application(s)

HPLC: suitable

L × I.D.

2 cm × 4 mm

matrix active group

amino phase

particle size

5 μm

Featured Industry

Food and Beverages

separation technique

normal phase

Looking for similar products? Visit Product Comparison Guide

Related Categories

Footnote

Kit includes one cartridge, a stand-alone holder, a piece of tubing, and 2 nuts and ferrules.

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC
Supelguard is a trademark of Sigma-Aldrich Co. LLC

Certificate of Analysis

Certificate of Origin

Sainath Babu et al.
Toxicology mechanisms and methods, 25(5), 410-416 (2015-05-30)
We have developed a simple, reversed-phase high-performance liquid chromatography (RP-HPLC) method for the determination of bisphenol A (BPA) in thermal paper cash register receipts (CRs). The method is suitable for analysis of other types of bisphenols and it involves an...
R Herráez-Hernández et al.
Journal of chromatographic science, 35(4), 169-175 (1997-04-01)
A chromatographic method for the analysis of amphetamine and related compounds in urine using 3,5-dinitrobenzoyl chloride (3,5-DNB) as a labeling reagent is presented. This assay is based on the employment of solid-phase extraction (SPE) cartridges for sample cleanup and derivatization....
F N Hussain et al.
Journal of chromatography. B, Biomedical sciences and applications, 716(1-2), 257-266 (1998-11-21)
We describe a sensitive high-performance liquid chromatographic method for the determination of 5-aminosalicylic acid and N-acetyl-5-aminosalicylic acid in rectal tissue biopsies. Samples were derivatised using propionic anhydride and proteins were precipitated with methanol. A Supelcosil ABZ column (150x4.6 mm I.D....
S Djabarouti et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 799(1), 165-172 (2003-12-09)
The aim of this study was to develop a specific and sensitive high-performance liquid chromatographic (HPLC) assay for the determination of levofloxacin in human plasma, bronchoalveolar lavage and bone tissues. The sample extraction was based on a fully automated liquid-solid...
E M Koves
Journal of chromatography. A, 692(1-2), 103-119 (1995-02-10)
A comprehensive approach to the analysis for many drugs in postmortem blood and biological fluids using high-performance liquid chromatography and diode array detection has been developed. To reduce the likelihood of co-eluting interference components of postmortem blood or other drugs...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service