Documents

59612

Supelco

SUPELCOSIL LC-18 Supelguard Cartridge

5 μm particle size, L × I.D. 2 cm × 2.1 mm

eCl@ss:
32110501
Pricing and availability is not currently available.

Quality Level

Agency/Method

suitable for USP L1

packaging

pkg of 1 kit

parameter

<70 °C temp. range

application(s)

HPLC: suitable

L × I.D.

2 cm × 2.1 mm

matrix

silica gel high purity, spherical

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

120 Å

pH range

2 - 7.5

Featured Industry

Food and Beverages

compatibility

use to protect LC-18, LC-PAH

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Related Categories

Footnote

Kit includes one cartridge, a stand-alone holder, a piece of tubing, 2 nuts and ferrules.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC
Supelguard is a trademark of Sigma-Aldrich Co. LLC
Certificate of Analysis
Certificate of Origin
Thadikamala Sathish et al.
Pakistan journal of pharmaceutical sciences, 22(1), 68-73 (2009-01-27)
An accurate, simple, reproducible and sensitive RP-HPLC method for the determination of bharangin has been developed and validated. The separation of bharangin and 2-nitroaniline (internal standard) was achieved on Supelcosil LC-18 (3micro, 150 x 4.6 mm i.d.) column using UV...
V Stocchi et al.
Analytical biochemistry, 146(1), 118-124 (1985-04-01)
A simple and rapid method for the determination of ATP, ADP, AMP, NADP+, NAD+, NADPH, and NADH in human erythrocytes is described. A single-step extraction procedure employing alkaline medium and CF 50A Amicon ultrafiltration membranes allows a simultaneous and total...
Mert Ilhan et al.
Evidence-based complementary and alternative medicine : eCAM, 2014, 578103-578103 (2014-08-29)
The aim of the present research is to evaluate the antiurolithiatic effect of the various extracts prepared from the fruits of Viburnum opulus L., in regard to its ethnobotanical record. To induce urolithiasis, 70 mg/kg sodium oxalate was injected to the...
L Marai et al.
Canadian journal of biochemistry and cell biology = Revue canadienne de biochimie et biologie cellulaire, 61(8), 840-849 (1983-08-01)
Natural triacylglycerols were resolved by high pressure liquid chromatography on Supelcosil LC-18 columns using a gradient of 30-90% propionitrile in acetonitrile as eluting solvent. The effluent was admitted to a quadrupole mass spectrometer via a direct liquid inlet interface. The...
Catrina L Dupard-Julien et al.
Analytical and bioanalytical chemistry, 387(3), 1027-1032 (2007-01-06)
A reversed-phase, high-performance liquid chromatography (RP-HPLC) method that allows quantitation of low levels of epoxides has been described. The method involved derivatization of epoxides using 100- to 1,000-fold excess N,N-diethyldithiocarbamate (DTC) at 60 degrees C for 20 min at neutral...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.