89374

Supelco

Heptadecafluorooctanesulfonic acid potassium salt

analytical standard

Synonym(s):
Potassium heptadecafluoro-1-octanesulfonate, Perfluorooctanesulfonic acid potassium salt
Linear Formula:
CF3(CF2)7SO3K
CAS Number:
Molecular Weight:
538.22
Beilstein/REAXYS Number:
3864579
MDL number:

grade

analytical standard

Quality Level

assay

≥92.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

277-280 °C (lit.)

SMILES string

[K+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C8HF17O3S.K/c9-1(10,3(13,14)5(17,18)7(21,22)23)2(11,12)4(15,16)6(19,20)8(24,25)29(26,27)28;/h(H,26,27,28);/q;+1/p-1

InChI key

WFRUBUQWJYMMRQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.