MilliporeSigma
All Photos(1)

Documents

91338

Supelco

Dimethyl 5-sulfoisophthalate sodium salt

analytical standard

Linear Formula:
NaO3SC6H3-1,3-(CO2CH3)2
CAS Number:
Molecular Weight:
296.23
MDL number:

grade

analytical standard

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

>300 °C (lit.)

SMILES string

[Na+].COC(=O)c1cc(cc(c1)S([O-])(=O)=O)C(=O)OC

InChI

1S/C10H10O7S.Na/c1-16-9(11)6-3-7(10(12)17-2)5-8(4-6)18(13,14)15;/h3-5H,1-2H3,(H,13,14,15);/q;+1/p-1

InChI key

LLHSEQCZSNZLRI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
524739136449234
mp

>300 °C (lit.)

mp

195 °C (dec.) (lit.)

mp

-

mp

−48 °C

assay

≥97.0% (HPLC)

assay

≥98.0% (HPLC)

assay

95-105% (T)

assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service