MilliporeSigma
All Photos(1)

97728-U

Supelco

Flash Silica

dried, pkg of 100 g, spherical

Synonym(s):
Silica gel
CAS Number:
EC Number:
MDL number:
NACRES:
NC.05

grade

high-purity grade

form

powder

packaging

pkg of 100 g

manufacturer/tradename

Sigma-Aldrich

technique(s)

LPLC: suitable

matrix

Silica

matrix active group

Silica

particle size

200-400 mesh
40-75 μm

pore size

0.7-0.9 cm3/g pore volume
70 Å pore size

surface area

480 m2/g

bp

2230 °C

mp

>1600 °C

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Compare Similar Items

View Full Comparison

1 of 4

This Item
23681022719697729-U
Flash Silica dried, pkg of 100 g, spherical

Supelco

97728-U

Flash Silica

Silica gel high-purity grade (Davisil Grade 643), pore size 150 Å, 200-425 mesh

Supelco

236810

Silica gel

Silica gel high-purity grade (9385), pore size 60 Å, 230-400 mesh particle size

Supelco

227196

Silica gel

Flash Silica dried, pkg of 1 kg, spherical

Supelco

97729-U

Flash Silica

form

powder

form

powder

form

powder

form

powder

packaging

pkg of 100 g

packaging

-

packaging

-

packaging

pkg of 1 kg

manufacturer/tradename

Sigma-Aldrich

manufacturer/tradename

Davisil®

manufacturer/tradename

EMD Millipore®

manufacturer/tradename

Sigma-Aldrich

technique(s)

LPLC: suitable

technique(s)

LPLC: suitable

technique(s)

LPLC: suitable

technique(s)

LPLC: suitable

matrix

Silica

matrix

Silica

matrix

Silica

matrix

Silica

matrix active group

Silica

matrix active group

silica

matrix active group

silica

matrix active group

Silica

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service