PS2063

Azocyclotin

analytical standard

Empirical Formula (Hill Notation):
C20H35N3Sn
CAS Number:
Molecular Weight:
436.22
Beilstein/REAXYS Number:
621636
EC Number:
MDL number:
PubChem Substance ID:

grade

analytical standard

form

neat

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-2063

application(s)

HPLC: suitable
gas chromatography (GC): suitable

Featured Industry

Agriculture
Environmental

format

neat

SMILES string

C1CCC(CC1)[Sn](C2CCCCC2)(C3CCCCC3)n4cncn4

InChI

1S/3C6H11.C2H2N3.Sn/c3*1-2-4-6-5-3-1;1-3-2-5-4-1;/h3*1H,2-6H2;1-2H;/q;;;-1;+1

InChI key

ONHBDDJJTDTLIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

RIDADR

UN 3146 6.1/PG 1

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.