Documents

ES5056

GreenSep PFP SFC 1.8 Micron HPLC Column

1.8 μm particle size, L × I.D. 10 cm × 2.1 mm

NACRES:
SB.52
Pricing and availability is not currently available.

Quality Level

mfr. no.

ES Industries, 522A91-GS-PFP

application(s)

HPLC: suitable

L × I.D.

10 cm × 2.1 mm

matrix active group

PFP (pentafluorophenyl) phase

particle size

1.8 μm

pore size

120 Å pore size

separation technique

supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

General description

Specifically designed for the separation of geometrical isomers as well as diastereomers. It is the column of choice in separating compounds that contain aromatic groups, polarizable electrons and conjugate systems. In addition, GreenSep PFP is useful for the separation of halogenated compounds. In many cases GreenSep PFP provides orthogonal separations when compared to GreenSep Nitro. GreenSep PFP is unsurpassed for its separation capabilities and is available in analytical columns, preparative columns and bulk media.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

GreenSep is a trademark of ES Industries
Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.