Documents

ES5212

GreenSep Ethyl Pyridine SFC 3 Micron HPLC Column

3 μm particle size, L × I.D. 10 cm × 4.6 mm

NACRES:
SB.52

Quality Level

manufacturer/tradename

ES Industries 125191-GS-PYE

application(s)

HPLC: suitable

L × I.D.

10 cm × 4.6 mm

matrix active group

2-ethyl pyridine phase

particle size

3 μm

pore size

120 Å pore size

separation technique

supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

General description

The type of chemicals separated on a GreenSep Ethyl Pyridine column would normally require the addition of an amine to the mobile phase as a peak shape modifier; however GreenSep Ethyl Pyridine does not require this addition. Ideal for chemicals that are functionalized with strong amine groups, GreenSep Ethyl Pyridine provides flexibility for the SFC chromatographer with mobile phase composition and fraction collection greatly simplified without the use of amino additives.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

GreenSep is a trademark of ES Industries

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.