Documents

ES5244

GreenSep Ethyl Pyridine II SFC 5 Micron HPLC Column

5 μm particle size, L × I.D. 15 cm × 4.6 mm

NACRES:
SB.52
Pricing and availability is not currently available.

Quality Level

mfr. no.

ES Industries, 135291-GS-PYE-II

application(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

matrix active group

2-ethyl pyridine phase

particle size

5 μm

pore size

120 Å pore size

separation technique

supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

General description

The column ideally suited for the retention and rapid separation of chemicals containing acidic groups. The chromatogram below shows the separation of chemicals that contain acidic groups and demonstrates the retention and separation capability that GreenSep Ethyl Pyridine II can deliver for the SFC chromatographer.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

GreenSep is a trademark of ES Industries
Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.