Documents

ES5333

GreenSep PFP SFC 5 Micron HPLC Column

5 μm particle size, L × I.D. 15 cm × 4.6 mm

NACRES:
SB.52

material

stainless steel column

Quality Level

product line

GreenSep

manufacturer/tradename

ES Industries 135291-GS-PFP

packaging

1 ea of

application(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

matrix

fully porous particle

matrix active group

PFP (pentafluorophenyl) phase

particle size

5 μm

pore size

120 Å pore size

operating pH

2-8

separation technique

supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

General description

Specifically designed for the separation of geometrical isomers as well as diastereomers. It is the column of choice in separating compounds that contain aromatic groups, polarizable electrons and conjugate systems. In addition, GreenSep PFP is useful for the separation of halogenated compounds. In many cases GreenSep PFP provides orthogonal separations when compared to GreenSep Nitro. GreenSep PFP is unsurpassed for its separation capabilities and is available in analytical columns, preparative columns and bulk media.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

GreenSep is a trademark of ES Industries

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service