MilliporeSigma
All Photos(1)

ES5442

ChromegaChiral Chiral CCC, 3 Micron HPLC Column

3 μm particle size, L × I.D. 10 cm × 3.0 mm

NACRES:
SB.52

material

stainless steel column

Quality Level

product line

ChromegaChiral

manufacturer/tradename

ES Industries 123151-CCC

packaging

1 ea of

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

10 cm × 3.0 mm

matrix

fully porous particle

matrix active group

3 chloro-4 methylphenylcarbamate and 3,5-dichlorophenylcarbamate phase

particle size

3 μm

pore size

1000 Å pore size

operating pH

2-8

separation technique

chiral
supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

ChromegaChiral CCC is a modified cellulose including the combination of 3-chloro-4 methylphenylcarbamate and 3,5-dichloro- phenylcarbamate bonding groups coated on high purity, high performance spherical silica particles. This combination of bonded groups stabilizes the solubility of coated phase making for a durable phase similar to other widely used coated phases. The use of cellulose modified with chlorinated phenyl groups provides for the separation for many previously unresolved/poorly resolved chiral mixtures by providing unique separation characteristics.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

ChromegaChiral is a trademark of ES Industries

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service