Documents

ES5611

ChromegaChiral Chiral CCS, 3 Micron HPLC Column

3 μm particle size, L × I.D. 15 cm × 4.6 mm

NACRES:
SB.52

manufacturer/tradename

ES Industries 135151-CCS

Quality Level

application(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

matrix active group

amylose, carbamate phase

particle size

3 μm

pore size

1000 Å pore size

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

General description

ChromegaChiral CCS (amylose tris [(S)-a-methylbenzylcarbamate]) permits the enantiomeric separation of 1-Indanol without the addition of DEA (Diethyl amine). Historically DEA has been commonly used to improve peak shape for chiral separations of compounds such as 1-Indanol. ChromegaChiral CCS separates the 1-Indanol enantiomers with sharp peaks without DEA, providing superior chiral separations, sample loading and peak shape performance.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

ChromegaChiral is a trademark of ES Industries

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.