K08970941

Kromasil® Guard Cartridge Starter Kit

phase C8 (octyl), use to protect 3.0 - 4.6 mm I.D. columns

eCl@ss:
32110501
NACRES:
SB.52
Pricing and availability is not currently available.

packaging

pkg of 1 kit

mfr. no.

Kromasil, KR100-5-C8-4.6XGSK

availability

available only in USA, Canada and Puerto Rico

application(s)

HPLC: suitable

L

10mm

matrix active group

C8 (octyl) phase

particle size

5 μm

pore size

100 Å

Featured Industry

Food and Beverages

compatibility

use to protect 3.0 - 4.6 mm I.D. columns

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

General description

All kits except K08970953 include 5 guard cartridges, guard cartridge holder, and coupler
K08970953 (Silica Starter Kit) includes 3 guard cartridges, guard cartridge holder, and coupler

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Kromasil is a registered trademark of EKA Chemicals AB
Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.