Sign In to View Organizational & Contract Pricing.
Select a Size
Pricing and availability is not currently available.
About This Item
Empirical Formula (Hill Notation):
C12H16O4
CAS Number:
Molecular Weight:
224.25
UNSPSC Code:
41116107
Skip To
Product Name
Senkyunolide I, phyproof® Reference Substance
grade
primary reference standard
product line
phyproof® Reference Substance
assay
≥90.0% (HPLC)
manufacturer/tradename
PhytoLab
technique(s)
HPLC: suitable
application(s)
food and beverages
pharmaceutical
format
neat
storage temp.
−20°C
Related Categories
General description
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Legal Information
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1B
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service