Skip to Content
MilliporeSigma

PHL86364

cis-2-Hydroxy-4-methoxycinnamic acid 2-glucoside

phyproof® Reference Substance

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
C16H20O9
CAS Number:
Molecular Weight:
356.32
UNSPSC Code:
41116107
NACRES:
NA.24

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

cis-2-Hydroxy-4-methoxycinnamic acid 2-glucoside, phyproof® Reference Substance

InChI

1S/C16H20O9/c1-23-9-4-2-8(3-5-12(18)19)10(6-9)24-16-15(22)14(21)13(20)11(7-17)25-16/h2-6,11,13-17,20-22H,7H2,1H3,(H,18,19)/b5-3-/t11-,13-,14+,15-,16-/m1/s1

InChI key

FQWZGEBZILOCET-QEQKPWIDSA-N

SMILES string

O[C@@H]([C@H](OC1=C(/C=C\C(O)=O)C=CC(OC)=C1)O2)[C@H]([C@@H]([C@H]2CO)O)O

grade

primary reference standard

product line

phyproof® Reference Substance

assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

format

neat

storage temp.

−20°C

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PHL83913PHL89850PHL83917
grade

primary reference standard

grade

primary reference standard

grade

primary reference standard

grade

primary reference standard

format

neat

format

neat

format

neat

format

neat

product line

phyproof® Reference Substance

product line

phyproof® Reference Substance

product line

phyproof® Reference Substance

product line

phyproof® Reference Substance

assay

≥90.0% (HPLC)

assay

≥95.0% (HPLC)

assay

≥95.0% (HPLC)

assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

manufacturer/tradename

PhytoLab

manufacturer/tradename

PhytoLab

manufacturer/tradename

PhytoLab

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service