Skip to Content
MilliporeSigma

PHL89676

Geraniol

phyproof® Reference Substance

Synonym(s):

(E)-Nerol, Geranyl alcohol, Lemonol, trans-3,7-Dimethyl-2,6-octadien-1-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2OH
CAS Number:
Molecular Weight:
154.25
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
1722456
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Geraniol, phyproof® Reference Substance

InChI

1S/C10H18O/c1-9(2)5-4-6-10(3)7-8-11/h5,7,11H,4,6,8H2,1-3H3/b10-7+

SMILES string

C\C(C)=C\CC\C(C)=C\CO

InChI key

GLZPCOQZEFWAFX-JXMROGBWSA-N

grade

primary reference standard

vapor density

5.31 (vs air)

product line

phyproof® Reference Substance

assay

≥90.0% (GC)

manufacturer/tradename

PhytoLab

technique(s)

HPLC: suitable

refractive index

n20/D 1.474 (lit.)

bp

229-230 °C (lit.)

density

0.879 g/mL at 20 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Packaging

This reference substance will be packed in micro-volume insert vials.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

flash_point_f

226.4 °F - closed cup

flash_point_c

108 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service