All Photos(1)

PS1000

Resmethrin

analytical standard

Empirical Formula (Hill Notation):
C22H26O3
CAS Number:
Molecular Weight:
338.44
Beilstein:
1626510
EC Number:
MDL number:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-1000

application(s)

agriculture
environmental

SMILES string

C\C(C)=C\C1C(C(=O)OCc2coc(Cc3ccccc3)c2)C1(C)C

InChI

1S/C22H26O3/c1-15(2)10-19-20(22(19,3)4)21(23)25-14-17-12-18(24-13-17)11-16-8-6-5-7-9-16/h5-10,12-13,19-20H,11,14H2,1-4H3

InChI key

VEMKTZHHVJILDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Peter Key et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 40(4), 585-594 (2005-07-29)
This study investigated the toxicity of various concentrations of technical resmethrin and Scourge on adult and larval Palaemonetes pugio, a common grass shrimp species. Two types of tests were conducted for each of the resmethrin formulations using adult and larval
Derek W Gammon
Pest management science, 63(7), 625-627 (2007-06-05)
West Nile virus is becoming increasingly prevalent in the USA, causing fever, encephalitis, meningitis and many fatalities. Spread of the disease is reduced by controlling the mosquito vectors by a variety of means, including the use of pyrethroid insecticides, which
Christopher J Williamson et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 44(8), 810-816 (2010-02-26)
The grass shrimp, Palaemonetes pugio, plays a large role in the marine ecosystem, serving as a vital link in the food web between many other species. Marine parasites such as the bopyrid isopod, Probopyrus pandalicola, reduce shrimp growth and reproductive
Marie E DeLorenzo et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 44(5), 455-460 (2010-02-26)
This study investigated the effects of increased temperature and salinity, two potential impacts of global climate change, on the toxicity of two common pesticides to the estuarine grass shrimp, Palaemonetes pugio. Larval and adult grass shrimp were exposed to the
Gary M Rand
Ecotoxicology (London, England), 11(2), 101-111 (2002-05-07)
A comparative aquatic hazard assessment of resmethrin was conducted to investigate the need for its restricted use classification by the US. EPA as an adult mosquito control agent. This paper describes the environmental fate and aquatic toxicity of resmethrin. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service