Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001307

Costunolide CRS

new

Synonym(s):

Costunolide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O2
CAS Number:
Molecular Weight:
232.32
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

description

(European Pharmacopoeia (EP) Reference Standard)

manufacturer/tradename

EDQM

application(s)

pharmaceutical

storage temp.

−20°C

SMILES string

C\C1=C/CC\C(C)=C\[C@H]2OC(=O)C(=C)[C@@H]2CC1

InChI

1S/C15H20O2/c1-10-5-4-6-11(2)9-14-13(8-7-10)12(3)15(16)17-14/h5,9,13-14H,3-4,6-8H2,1-2H3/b10-5+,11-9+/t13-,14+/m0/s1

InChI key

HRYLQFBHBWLLLL-AHNJNIBGSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Costunolide is a potent inducer of apoptosis and potent anti-cancer agent. Costunolide has anti-inflammatory, anti-viral, anti-fungal, antimycobacterial activity and inhibits telomerase activity.
Costunolide is a sesquiterpene lactone originally isolated from plants widely used in many herbal medicines. It has a variety of effects. Costunolide is a potent inducer of apoptosis, a potent anti-cancer agent, has anti-inflammatory, anti-viral, anti-fungal, antimycobacterial activity and has been shown to inhibit telomerase activity.

Packaging

Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit EDQM reference substance catalogue.

Other Notes

EP issued certificate and SDS can be found here.
Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service