Skip to Content
MilliporeSigma

Y0001810

Mycophenolate sodium CRS

new

Synonym(s):

Mycophenolic acid monosodium salt, Sodium mycophenolate

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C17H19NaO6
CAS Number:
Molecular Weight:
342.32
UNSPSC Code:
41116107

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

pharmaceutical primary standard

description

(European Pharmacopoeia (EP) Reference Standard)

manufacturer/tradename

EDQM

application(s)

pharmaceutical

storage temp.

2-8°C

InChI

1S/C17H20O6.Na/c1-9(5-7-13(18)19)4-6-11-15(20)14-12(8-23-17(14)21)10(2)16(11)22-3;/h4,20H,5-8H2,1-3H3,(H,18,19);/q;+1/p-1/b9-4+;

InChI key

DOZYTHNHLLSNIK-JOKMOOFLSA-M

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit EDQM reference substance catalogue.

Other Notes

EP issued certificate and SDS can be found here.
Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service