Y0002350
Pulchinenoside B4 CRS

Synonym(s):
Anemoside B4, Chinensioside A, Pulchinenoside B4
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C59H96O26
CAS Number:
Molecular Weight:
1221.38
MDL number:
UNSPSC Code:
41116107
Skip To
Pricing and availability is not currently available.
grade
pharmaceutical primary standard
description
(European Pharmacopoeia (EP) Reference Standard)
manufacturer/tradename
EDQM
application(s)
pharmaceutical
storage temp.
2-8°C
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Packaging
Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit EDQM reference substance catalogue.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service