Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0002424

Lercanidipine hydrochloride CRS

new

Synonym(s):

Lercanidipine hydrochloride, 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-2-[(3,3-diphenylpropyl)methylamino]-1,1-dimethylethyl methyl ester 3,5-pyridinedicarboxylic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H41N3O6 · HCl
CAS Number:
Molecular Weight:
648.19
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

description

(European Pharmacopoeia (EP) Reference Standard)

manufacturer/tradename

EDQM

application(s)

pharmaceutical

storage temp.

2-8°C

SMILES string

O=C(OC)C1=C(C)NC(C)=C(C(OC(C)(C)CN(C)CCC(C2=CC=CC=C2)C3=CC=CC=C3)=O)C1C4=CC([N+]([O-])=O)=CC=C4.Cl

InChI

1S/C36H41N3O6.ClH/c1-24-31(34(40)44-6)33(28-18-13-19-29(22-28)39(42)43)32(25(2)37-24)35(41)45-36(3,4)23-38(5)21-20-30(26-14-9-7-10-15-26)27-16-11-8-12-17-27;/h7-19,22,30,33,37H,20-21,23H2,1-6H3;1H

InChI key

WMFYOYKPJLRMJI-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Lercanidipine hydrochloride is a L-type (Cav1.2b) vascular channel antagonist; L-type (Cav1.2a) cardiac channel agonist voltage-dependent and highly lipophylic compound, which exhibits a slower onset and longer duration of action than other calcium channel antagonists; an antihypertensive agent in patients with mild-to-moderate hypertension; more vasoselective than lacidipine and amlodipine.
Lercanidipine hydrochloride is a L-type (Cav1.2b) vascular channel antagonist; L-type (Cav1.2a) cardiac channel agonist.

Packaging

Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit EDQM reference substance catalogue.

Other Notes

EP issued certificate and SDS can be found here.
Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service