Skip to Content
MilliporeSigma

Y0002432

Rutoside for system suitability CRS

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Rutin trihydrate, Quercetin-3-rutinoside trihydrate, Vitamin P trihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
C27H30O16 · 3H2O
CAS Number:
Molecular Weight:
664.56
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
75455
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Rutoside for system suitability CRS, European Pharmacopoeia (EP) Reference Standard

InChI key

NLLBWFFSGHKUSY-JPRRWYCFSA-N

SMILES string

O.O.O.C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(Oc4cc(O)cc(O)c4C3=O)c5ccc(O)c(O)c5)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H30O16.3H2O/c1-8-17(32)20(35)22(37)26(40-8)39-7-15-18(33)21(36)23(38)27(42-15)43-25-19(34)16-13(31)5-10(28)6-14(16)41-24(25)9-2-3-11(29)12(30)4-9;;;/h2-6,8,15,17-18,20-23,26-33,35-38H,7H2,1H3;3*1H2/t8-,15+,17-,18+,20+,21-,22+,23+,26+,27-;;;/m0.../s1

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

application(s)

pharmaceutical

storage temp.

2-8°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Other Notes

EP issued certificate and SDS can be found here.
Sales restrictions may apply.

Packaging

Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit EDQM reference substance catalogue.

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service