Skip to Content
MilliporeSigma

SY3390162173

Synthonix Corporation

Morpholin-3-one

Synonym(s):

3-morpholinone

Sign In to View Organizational & Contract Pricing.

Select a Size

1 G

$26.45

25 G

$33.35

50 G

$54.05

Vendor SKU: M7700-1G

$26.45


Ships in 10 days - from Aldrich Partner.


About This Item

Linear Formula:
C4H7O2N1
CAS Number:
Molecular Weight:
101.11
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C4H7NO2/c6-4-3-7-2-1-5-4/h1-3H2,(H,5,6)

Inchi Key

VSEAAEQOQBMPQF-UHFFFAOYSA-N

Shipping Temperature

Room Temperature

Purity

97%

Storage temp.

Room Temperature

Country of Origin

US

Physical Form

Solid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service