MilliporeSigma
All Photos(1)

SY3H3D6755B5

Synthonix Corporation

2-Amino-5-bromobenzonitrile

CAS Number:
Molecular Weight:
197.03
MDL number:

Iupac Name

2-amino-5-bromobenzonitrile

Inchi Code

1S/C7H5BrN2/c8-6-1-2-7(10)5(3-6)4-9/h1-3H,10H2

InChI key

OATYCBHROMXWJO-UHFFFAOYSA-N

storage temp.

Refrigerator

Purity

97%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Crystal - Powder / Colour: White - Very pale yellow

Product Link

https://synthonix.com/39351.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://synthonix.com/39351.html

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service